Would you like to log in again?
KidWind simpleGEN AC Generator
Supplier: VERNIER SOFTWARE & TECH SE
KW-SGEN
470165-156EA 78.99 USD InStock
470165-156
KidWind simpleGEN AC Generator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The simpleGEN is an easy-to-build AC generator that allows students to explore the basics of electrical generator design. With the simpleGEN, you can demonstrate Faraday’s law, light LEDs, and perform experiments that explore how coils, magnets, and rotation affect electrical generation. With some quick modifications, you will be able to convert your generator to a simple motor and explore additional variables. You can also take your experiments to the next level by converting your simpleGEN into a wind turbine nacelle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty