Would you like to log in again?
Solar Thermal Exploration Kit
Supplier: VERNIER SOFTWARE & TECH SE
KW-STXK
470165-162EA 63.99 USD InStock
470165-162
Solar Thermal Exploration Kit
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
The Solar Thermal Exploration Kit is modeled after a domestic solar water-heating system. Explore variables such as box color, light intensity, tube design, rate of water pumping, and amount of time it takes to heat the water. Learn about what makes a solar water heating system more or less efficient.
Solar water heaters are becoming a popular alternative to conventional water heaters because they save the homeowner money, and they reduce carbon emissions. The instructions that come with these kits include some basic information on solar water heating systems and how they work.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty