Would you like to log in again?
Human Recombinant Cardiotrophin 1 (from E. coli)
Supplier: BioVision
AB285453-1MG AB285453-50UG AB285453-10UG
10190-170EA 1846.27 USD InStock
10190-170 10190-166 10190-168
Human Recombinant Cardiotrophin 1 (from E. coli)
Proteins and Peptides
>98% Pure human recombinant Cardiotrophin-1.
Human Cardiotrophin-1 (CT-1) is a member of the leukemia inhibitory/ciliary neurotrophic factor/Oncostatin M/interleukin-6/interleukin-11 family of cytokines. Cardiotrophin-1 affects cells through interaction with the gp 130 receptor subunit. Recombinant human Cardiotrophin-1 is a 22.5 kDa protein containing 200 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty