Would you like to log in again?
KidWind Wind Turbine Hub
Supplier: VERNIER SOFTWARE & TECH SE
KW-WTH3 KW-WTH10
470165-168PK 21.99 USD InStock
470165-168 470165-166
KidWind Wind Turbine Hub
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The hub will press fit onto the shaft of many small DC generators; just attach your own blades and begin experimenting with wind power! The hub is designed to allow you to change blade angles and/or interchange blade sets quickly and easily. An improved stainless-steel insert can take a beating in your classroom year after year without failing!
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty