Would you like to log in again?
Renewable Energy with Vernier Lab Book
Supplier: VERNIER SOFTWARE & TECH SE
REV
470165-170EA 51.99 USD InStock
470165-170
Renewable Energy with Vernier Lab Book
Educational Materials Physics Educational Materials Physics Textbooks
The Renewable Energy with Vernier lab book features 26 experiments in wind and solar energy.
  • Aligned to NGSS
  • Pre and post laboratory assessments
  • Linked to a real world scenario
The lab book contains a combination of explorations, traditional experiments, inquiry investigations, engineering projects, and more. In these experiments, students learn about renewable energy, experiment with variables affecting power output from model wind turbines and solar panels, and participate in engineering projects such as building a wind farm or a solar charger to meet certain design criteria. Aligned with the Next Generation Science Standards, NGSS, all of the experiments are designed to support students in three-dimensional learning with Disciplinary Core Ideas, Science and Engineering Practices, and Crosscutting Concepts identified for each experiment within the Instructor pages.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty