Would you like to log in again?
L-JNK Inhibitor I Peptide
Supplier: BioVision
AB52324-50
10190-036EA 177.06 USD InStock
10190-036
L-JNK Inhibitor I Peptide
Proteins and Peptides
L-JNK Peptide Inhibitor competitively blocks the interaction between JNK and c-Jun, thereby inhibiting the signaling events downstream of JNK, like c-Jun, ATF-2 and ELK1 phosphorylation. To convert JIP-1/IB-1 into cell permeable inhibitors of JNK (JNKI1) the minimal 20 amino acids inhibitory sequence of JIP-1/IB1 was covalently linked to the 10 amino acids recognized by TAT transporter. The L-JNKI1 is a potent inhibitor that is specific for JNK and can be used for in vitro applications.
L-JNK Peptide Inhibitor competitively blocks the interaction between JNK and c-Jun, thereby inhibiting the signaling events downstream of JNK, like c-Jun, ATF-2 and ELK1 phosphorylation. To convert JIP-1/IB-1 into cell permeable inhibitors of JNK (JNKI1) the minimal 20 amino acids inhibitory sequence of JIP-1/IB1 was covalently linked to the 10 amino acids recognized by TAT transporter. The L-JNKI1 is a potent inhibitor that is specific for JNK and can be used for in vitro applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty