Would you like to log in again?
Human Recombinant FABP3 (from E. coli)
Supplier: BioVision
AB285573-25UG AB285573-1MG AB285573-100UG
10190-444EA 1026.57 USD InStock
10190-444 10190-446 10190-448
Human Recombinant FABP3 (from E. coli)
Proteins and Peptides
>98% Pure Recombinant human FABP3.
Test in process
Human Fatty Acid Binding Protein-3 (FABP-3) exhibits high affinity for small lipophilic ligands. Studies suggest that FABPs are involved in the uptake and metabolism of fatty acids, maintenance of Cellular membrane fatty acids levels, intraCellular trafficking, modulation of specific enzymes of lipid metabolic pathways, as well as in the modulation of cell growth and differentiation
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty