Would you like to log in again?
Human Recombinant FGF19 (from E. coli)
Supplier: BioVision
AB285586-1MG AB285586-25UG AB285586-100UG
10190-478EA 566.91 USD InStock
10190-478 10190-474 10190-476
Human Recombinant FGF19 (from E. coli)
Proteins and Peptides
>95% Pure active Recombinant human FGF-19.
Determined by a cell proliferation assay using balb/c 3T3 cells. The expected ED₅₀ for this effect is 50-200 ng/ml.
Fibroblast Growth Factor-19 (FGF-19) is a high affinity heparin dependent ligand for FGFR4. Recombinant human FGF-19 is an 21.8 kDa protein containing 195 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty