Would you like to log in again?
Mouse Recombinant Rank (from E. coli)
Supplier: BioVision
AB51941-10
10191-434EA 249.2 USD InStock
10191-434
Mouse Recombinant Rank (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant murine sRANKL.
Measured by its ability to induce osteoclast formation in RAW264.7 cells using a concentration of 5-10 ng/ml.
Murine RANK (receptor activator of NF-kB) is a newly discovered member of the TNFR family, identified as a dendritic cell membrane protein. The recombinant murine sRANK Ligand is a soluble 19.4 kDa protein, comprising the 174 amino acid residue TNF homologous region of RANKL.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty