Would you like to log in again?
Human Recombinant Tace (from Insect Cells)
Supplier: BioVision
AB138784-20UG AB138784-100UG AB138784-1MG
10191-448EA 2707.79 USD InStock
10191-448 10192-046 10190-872
Human Recombinant Tace (from Insect Cells)
Proteins and Peptides
>95% Pure active Recombinant Human TACE.
Activity of TACE was tested using the synthetic peptide, Abz-Leu-Ala-Gln-Ala-Val-Arg-Ser-Ser-Ser-Arg-Dpa(Dnp)-NH2, as a substrate.
TACE (TNF-alpha converting enzyme, ADAM17) is a membrane-bound Metalloprotease from the ADAM (a disintegrin and Metalloprotease) family of proteins. It cleaves 26kDa type II membrane-bound precursor of TNF-a into 17 kDa mature form which is secreted. TACE is also known to process other membrane proteins including TNF receptor, TGF-alpha and amyloid protein precursor. Recombinant active untagged TACE amino acid sequence is from Arg215 to Asn651. The apparent molecular weight is bigger than the calculated MW due to the protein posttranslational modification in eukaryotic system.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty