Would you like to log in again?
Human Recombinant BMP12 (from E. coli)
Supplier: BioVision
AB285592-100UG AB285592-1MG AB285592-20UG
10190-504EA 1325.31 USD InStock
10190-504 10190-506 10190-508
Human Recombinant BMP12 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant human BMP-12/GDF-7.
The protein is biologically active as determined by its ability to induce alkaline phosphatase production by ADTC5 mouse chondrogenic cells. The ED₅₀ for this effect is typically 0.2 – 2.5 µg/ml.
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. BMP-12 is highly conserved across species. BMP-12 regulates chondrogenesis, bone morphogenesis, and neuron differentiation. Recombinant human BMP-12 is a 27 kDa disulfide-linked homodimer containing two 122 amino acid polypeptide chains.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty