Would you like to log in again?
Human Recombinant Cyan Fluorescent1 (from E. coli)
Supplier: BioVision
AB285703-5MG AB285703-1MG AB285703-100UG
10190-830EA 850.6 USD InStock
10190-830 10190-832 10190-834
Human Recombinant Cyan Fluorescent1 (from E. coli)
Proteins and Peptides
>97% Pure Recombinant Cyan Fluorescent Protein
The recombinant CFP (Cyan Fluorescent Protein) is expressed and purified from transformed E. coli using a method that ensures high purity and maximal CFP fluorescence. Endotoxin has been removed, so the protein is suitable for in vivo injection or cell culture applications. The protein is a 31.3 kDa monomer with 284 amino acids and an N-terminal His-tag, Ex./Em. = 458/480 nm, extinction coefficient 26000M-1CM-1.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty