Would you like to log in again?
Human Recombinant C5A (from E. coli)
Supplier: BioVision
AB61918-20
10190-828EA 282.07 USD InStock
10190-828
Human Recombinant C5A (from E. coli)
Proteins and Peptides
>95% Pure active Recombinant Human C5a
Human recombinant C5a is as active as that of serum derived C5a based on enzyme release assay of myeloperoxidase. The current lot of ED₅₀ = 1.36 nmolar.
Human complement C5a is a 74-amino acid glycopeptide generated during complement activation from the α-chain of complement C5. C5a is a potent chemotactic factor for human peripheral blood neutrophils and monocytes, and is believed to play an important role in a number of inflammatory conditions. The 74 amino acid, human C5a protein expressed in E. coli has an activity similar to that of serum derived C5a based on enzyme assay of myeloperoxidase release.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty