Would you like to log in again?
Human Recombinant BMP11 (from E. coli)
Supplier: BioVision
AB285594-1MG AB285594-10UG AB285594-50UG
10190-510EA 467.32 USD InStock
10190-510 10190-512 10190-514
Human Recombinant BMP11 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant human BMP-11.
The protein is biologically active as determined by its ability to induce alkaline phosphatase production by C2C12 cells. The ED₅₀ is in the range of 0.4-4 µg/ml.
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. Recombinant human BMP-11 is N-terminus His-tagged protein with a molecular weight of 32.6 kDa (for homodimeric) and 16.2 (for monomeric) consisting of 136 amino acid polypeptide chains.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty