Would you like to log in again?
KingFisher™ Duo Prime Purification System, Thermo Scientific
Supplier: Thermo Scientific
5400110 5400110 N16640 N16640
10755-330EA 36927.17 USD InStock
10755-330 CA10755-330 10755-332 CA10755-332
KingFisher™ Duo Prime Purification System, Thermo Scientific
Reap high yields of quality nucleic acids from a variety of starting materials with the Thermo Scientific KingFisher Duo Prime purification system
KingFisher Duo Prime Purification System can isolate DNA, RNA and proteins from a variety of starting materials, including cell-free body fluids, blood, bacteria, cell cultures, tissue and plant samples; with volumes ranging up to 5 mL. All KingFisher systems use automated magnetic bead technology, which allows excellent recovery of samples with great reproducability and efficiency. Built-in UV lamp delivers easy and effective decontamination. Optional bar-code reader allows users to track samples directly in the internal software. Use with optimized KingFisher Pure DNA and RNA isolation kits and protocols for a complete, easy-to-use solution, or customize with Thermo Scientific BindIt™ software to create your own protocols.
Recommended for DNA and RNA isolation from various starting materials, proteomic applications and cell isolation. Protocols can be modified or developed, as well as imported directly to the instrument using PC or via USB stick. BindIt software enables you to save run log files individually, providing traceability and complete data management.
This compact instrument is ideal for laboratories requiring nucleic acid purification, and is easily modified for use in proteomics and cell isolation. It provides excellent reproducibility with minimal hands-on time, and has a small footprint in the lab. New features, including a UV lamp for easy decontamination and optional bar-code reader enhance functionality.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty