Would you like to log in again?
Human Recombinant IFN alpha 1 (from E. coli)
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB285612-1MG AB285612-100UG AB285612-10UG
10190-554EA 558.28 USD InStock
10190-554 10190-556 10190-552
Human Recombinant IFN alpha 1 (from E. coli)
Proteins and Peptides
>95% Pure active Recombinant human IFN-alpha 1.
Recombinant human IFN-α1 is fully biologically active. The specific activity was determined to be 1 x 10⁸ units/mg.
IFN-α1 is a lymphoid factor that possesses potent anti-viral activity. Human IFN-a1 is a 19.3 kDa protein containing 166 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty