Would you like to log in again?
Human Recombinant VEGFC (from E. coli)
Supplier: BioVision
AB51968-10 AB51968-50 AB51968-1
10191-442EA 649.21 USD InStock
10191-442 10191-440 10191-438
Human Recombinant VEGFC (from E. coli)
Proteins and Peptides
>90% Pure active Recombinant human VEGF-C.
Activity of VEGF-C is tested by the stimulation of VEGF-R3 phosphorylation in cells transfected with VEGF-R3. The expected ED₅₀ is >200 ng/ml.
The recombinant VEGF-C (Vascular Endothelial Growth Factor-C) is a 125 amino acid protein that lacks the N-terminal signal peptide (from T103 to R227 of Gene Bank U58111). Human VEGF-C exhibits about 85% homology with murine VEGF-C.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty