Would you like to log in again?
Mouse Recombinant Adiponectin (from E. coli)
Supplier: BioVision
AB285685-100UG AB285685-1MG AB285685-25UG
10190-746EA 1528.64 USD InStock
10190-746 10190-748 10190-750
Mouse Recombinant Adiponectin (from E. coli)
Proteins and Peptides
>90% Pure Recombinant murine Adiponectin.
Adiponectin (also called Acrp30, AdipoQ) is an adipocyte specific secreted protein that circulates in the plasma. It is induced during adipocyte differentiation and it secretion is stimulated by insulin. Mouse adiponectin shares about 83% amino acid identity with that of human. Adiponectin plays a role in various physiological processes such as energy homeostasis and obesity. Plasma levels of adiponectin are reduced in obese humans, and decreased levels are associated with insulin resistance and hyperinsulinemia.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty