Would you like to log in again?
Mouse Recombinant C5A (from E. coli), HIS Tag
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB285676-1MG AB285676-100UG AB285676-20UG
10190-730EA 1754.35 USD InStock
10190-730 10190-732 10190-734
Mouse Recombinant C5A (from E. coli), HIS Tag
Proteins and Peptides
>95% Pure active Recombinant murine C5a (His-Tag).
ED₅₀ 1-3 nM as determined by enzyme release assay of myeloperoxidase.
Mouse complement C5a is a 77-amino acid peptide generated during complement activation from the α-chain of complement C5. Mouse C5a shares 60% and 82% amino acid sequence identity to human and rat C5a, respectively. C5a binds to a signaling G-protein coupled receptor (GPCR) (C5aR/CD88), inducing neutrophil chemotaxis and endothelial cell activation. It also triggers an oxidative burst in macrophages and neutrophils, and induces release of histamine in basophils and mast cells. The 9 kDa mouse C5a protein expressed in E. coli contains a N-terminal His-Tag sequence.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty