Would you like to log in again?
Human Recombinant Noggin (from E. coli)
Supplier: BioVision
AB285619-100UG AB285619-20UG AB285619-1MG
10190-590EA 1149.13 USD InStock
10190-590 10190-592 10190-594
Human Recombinant Noggin (from E. coli)
Proteins and Peptides
>95% Pure active Recombinant human Noggin.
Determined by its ability to inhibit 5 ng/ml of BMP-4-induced alkaline phosphatase production by ATDC chondrogenic cells. The expected ED₅₀ for the effect is 0.05-0.08 µg/ml of Noggin
Noggin belongs to a group of diffusible proteins which bind to ligands of the TGF-β family and regulate their activity by inhibiting their access to signaling receptors. Noggin was originally identified as a BMP-4 antagonist whose action is critical for proper formation of the head and other dorsal structures. Consequently, Noggin has been shown to modulate the activities of other BMPs including BMP-2,-7,-13, and -14. Targeted deletion of Noggin in mice results in prenatal death and recessive phenotype displaying a severely malformed skeletal system. Conversely, transgenic mice over-expressing Noggin in mature osteoblasts display impaired osteoblastic differentiation, reduced bone formation, and severe osteoporosis. Recombinant human Noggin is a 23.1 kDa non-disulfide-linked homodimer consisting of a total of 206 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty