Would you like to log in again?
Human Recombinant Heat Shock 90 (from Insect cells (Sf9))
Supplier: BioVision
AB52154-25
10190-700EA 399.9 USD InStock
10190-700
Human Recombinant Heat Shock 90 (from Insect cells (Sf9))
Proteins and Peptides
>99% Pure Recombinant human Heat Shock Protein 90.
Not available at this time
Hsp90 is a molecular chaperone and is one of the most abundant proteins in unstressed cells. In mammalian cells, there are two genes encoding cytosolic Hsp90 homologues, with the human Hsp90α showing 85% sequence identity to Hsp90β. The recombinant Hsp90 produced from Sf9 cells represents the full length (a.a. 1-723) of Hsp90b subunit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty