Would you like to log in again?
CT-1000HT Series Constant Temperature Baths, 120 V, Cannon Instrument
Supplier: Cannon Instrument Company
9726-A27
47735-070EA 15644.32 USD InStock
47735-070
CT-1000HT Series Constant Temperature Baths, 120 V, Cannon Instrument
Baths Viscometer Baths
This durable and accurate insulated bath offers quick-set temperature control options and ASTM D 445 reliability
Up to seven viscometer tubes/holders can be placed in the bath. The unit is equipped with several safety features, including a bath level float and over-temperature thermistor.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty