Would you like to log in again?
Human Recombinant CTGF (from E. coli)
Supplier: BioVision
AB285639-100UG AB285639-1MG AB285639-20UG
10191-968EA 612.86 USD InStock
10191-968 10191-966 10191-964
Human Recombinant CTGF (from E. coli)
Proteins and Peptides
>95% Pure active Recombinant human CTGF.
ED₅₀ 1.0–2.0 μg/ml determined by the dose-dependent stimulation of the proliferation of HUVEC cells.
CTGF is a member of the CCN family of secreted cysteine rich regulatory proteins and is the major mitogenic and chemoattractant protein produced by μmbilical vein and Vascular endothelial cells. CTGF stimulates the proliferation and differentiation of chondrocytes, induces angiogenesis, promotes cell adhesion of fibroblasts, endothelial, and epithelial cells, and binds to IGF, TGF β1, and BMP-4. Cell migration and adhesion are signaled through binding to specific cell surface integrins and to heparin sulfate proteoglycans CTGF (98 a.a.), a lower molecular weight isoform containing the C-terminal portion of the full length CTGF protein, exerts full heparin binding, cell adhesion, and mitogenic CTGF activity. Recombinant human CTGF is a 11.2 kDa protein of 98 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty