Would you like to log in again?
Cannon Bath Cooler, 115 V, Cannon Instrument Company
Supplier: Cannon Instrument Company
9726-D05
47735-078EA 7153.28 USD InStock
47735-078
Cannon Bath Cooler, 115 V, Cannon Instrument Company
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The compact and efficient Cannon Bath Cooler (CBC) uses an advanced technological design to maintain a liquid bath below room temperature
With the CBC, bath temperatures can be lowered up to 15°C below ambient. The CBC consists of a thermoelectric Cooling Unit and a separate power supply.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty