Would you like to log in again?
Bovine Native Aprotinin1 (from Lung)
Supplier: BioVision
AB49828-5
10190-618EA 124.41 USD InStock
10190-618
Bovine Native Aprotinin1 (from Lung)
Proteins and Peptides
>98% Pure active Aprotinin.
5 x 10⁶ IU/mg.
Aprotinin inhibits the activity of several proteolytic enzymes such as chymotrypsin, kallikrein, plasmin and trypsin. It is present in blood and in most tissues, with a high concentration in lung, inhibits pro-inflammatory cytokine release and maintains glycoprotein homeostasis. In platelets, aprotinin reduces glycoprotein loss (e.g., GpIb, GpIIb/IIIa), while in granulocytes it prevents the expression of pro-inflammatory adhesive glycoproteins. Aprotinin is a natural proteinase inhibitor polypeptide consisting of fifty-eight amino acids arranged in a single polypeptide chain, cross-linked by three disulfide bridges and having a molecular mass of 6512 Daltons. Aprotinin is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty