Would you like to log in again?
Rat Recombinant Prolactin (from E. coli)
Supplier: BioVision
AB285629-10UG AB285629-50UG AB285629-1MG
10190-610EA 4213.46 USD InStock
10190-610 10190-612 10190-608
Rat Recombinant Prolactin (from E. coli)
Proteins and Peptides
>95% Pure active Recombinant rat Prolactin.
The ED₅₀ determined by the dose-dependent stimulation of the proliferation of rat lymphoma, Nb2-11 cells was found to be in the range of 0.1-1.0 ng/ml.
Prolactin (also known as mammotrophin, luterotropic hormone, and lutetropin) is a neuroendocrine hormone secreted by the pituitary gland. Its primary function is to promote and maintain lactation during pregnancy and suckling. In addition, Prolactin plays an immune-regulatory role by stimulating the activities of ornithine decarboxylase and protein kinase C, which are important for the proliferation, differentiation, and function of lymphocytes. Recombinant murine Prolactin is a 22.5 kDa globular protein containing 198 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty