Would you like to log in again?
Normal Donkey Serum, ChromPure, Jackson Immunoresearch
Supplier: Jackson Immunoresearch Laboratories
ChromPure
017-000-121 017-000-001
102644-006EA 78.77 USD InStock
102644-006 102643-998
Normal Donkey Serum, ChromPure, Jackson Immunoresearch
Sera (Serum)
Normal serums are lipid extracted to improve clarity.
  • Dialyzed against PBS containing sodium azide
  • Freeze-dried
  • Strongly recommended blocking agent
Normal serums are lipid extracted to improve clarity, dialyzed against phosphate buffered saline (PBS) containing sodium azide, and freeze-dried. Normal serum diluted to 5% (v/v) in PBS is strongly recommended as a blocking reagent to reduce background from non-specific, conserved sequence and/or Fc receptor binding. Best results are obtained with diluted normal serum from the same host as the labeled antibody, as a separate incubation step before addition of the primary antibody.
Packaging: Glass Ampuole
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty