Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Streptavidin Antibody (HRP (Horseradish Peroxidase))

Supplier: Cytiva
RPN4401 RPN4401
95019-224EA 1917.74 USD InStock
CA95019-224L 95019-224
Anti-Streptavidin Antibody (HRP (Horseradish Peroxidase))
Antibodies
For use in ELISA systems. For ELISA, it is recommended that the conjugate should be diluted in the range 1:4000 to 1:8000.

Amdex conjugates are already supplied as components of the Biotrak high-sensitivity cytokine ELISAs, complete with fully-optimized protocols. Easily soluble in cold and hot water.

Type:
Antigen: Streptavidin
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty