Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD10 Mouse Monoclonal Antibody [clone: 56C6]

Supplier: Diagnostic BioSystems

PDM107 PDM107
99990-990EA 340.33 USD InStock
99990-990 CA99990-990
Anti-CD10 Mouse Monoclonal Antibody [clone: 56C6]
Antibodies
Type:
Antigen: CD10
Clonality: Monoclonal
Clone: 56C6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty