Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GLB1 Mouse Monoclonal Antibody

Supplier: Promega
Z3781
PAZ3781EA 66.15 USD InStock
PAZ3781
Anti-GLB1 Mouse Monoclonal Antibody
Antibodies
This antibody [subclass IgG(2a)(kappa)] recognizes E. coli beta-galactosidase.

  • Monoclonal Antibody that Recognizes E. coli β-galactosidase
  • Antibody [subclass IgG2a(κ)] was purified from ascites of a mouse hybridoma
  • Can be used for several immunological assays
  • Supplied at a concentration of 2.0-2.5mg/ml

This antibody [subclass IgG(2a)(kappa)] was purified from ascites of a mouse hybridoma and recognizes E. coli beta-galactosidase.

Type:
Antigen: ß-Galactosidase
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty