Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TUBB3 Mouse Monoclonal Antibody [clone: 5G8]

Supplier: Promega
G7121
PAG7121EA 565.7 USD InStock
PAG7121
Anti-TUBB3 Mouse Monoclonal Antibody [clone: 5G8]
Antibodies
Anti-betaIII Tubulin mAb is a protein G-purified IgG(1) monoclonal antibody against a peptide (EAQGPK) corresponding to the C-terminus of betaIII tubulin.

  • Neuron-Specific Monoclonal Antibody That Reacts with C-Terminal βIII Tubulin
  • Antigenic peptide corresponding to the C terminus (EAQGPK) of βIII tubulin
  • Mouse monoclonal IgG1 (clone 5G8) supplied as 1mg/ml in PBS containing no preservatives
  • Cross-reacts with most mammalian species. Does not label nonneuronal cells (e.g., astrocytes).

Anti-betaIII Tubulin mAb is a protein G-purified IgG(1) monoclonal antibody (from clone 5G8) raised in mice against a peptide (EAQGPK) corresponding to the C-terminus of betaIII tubulin. It is directed against betaIII tubulin, a specific marker for neurons. The major use of this antibody is for labeling neurons in tissue sections and cell culture. The antibody has been tested to perform in frozen and paraffin-embedded sections of rat brain, cerebellum and spinal cord, human and rat fetal CNS progenitor cell cultures and adult human paraffin-embedded brain.

Delivery information: Mouse monoclonal IgG1 (clone 5G8) supplied as 1 mg/ml in PBS containing no preservative.

Type:
Antigen: TUBB3
Clonality: Monoclonal
Clone: 5G8
Conjugation:
Epitope: C-terminus
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty