Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-POGZ Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-A89 600-401-A89
RL600401A89EA 518 USD InStock
RL600401A89 CA600401A89
Anti-POGZ Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: POGZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype:
Reactivity: Human, Dog, Short-Tailed Opossum, Bovine, Rat, Chimpanzee, Macaque, Olive Baboon, Chicken

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty