Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA1B Mouse Monoclonal Antibody [clone: C92F3A-5]

Supplier: Rockland Immunochemicals
200-301-A27 200-301-A27
RL200-301-A27EA 528 USD InStock
RL200-301-A27 CA200-301-A27
Anti-HSPA1B Mouse Monoclonal Antibody [clone: C92F3A-5]
Antibodies
Type: Primary
Antigen: HSPA1B
Clonality: Monoclonal
Clone: C92F3A-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, C.elegans, Chicken, Dog, Drosophila, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, carp

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty