Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: 15A3]

Supplier: Rockland Immunochemicals
200-301-A98 200-301-A98
RL200-301-A98EA 528 USD InStock
CA200-301-A98 RL200-301-A98
Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: 15A3]
Antibodies
Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: 15A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty