Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-A3GALNT, A3GALT1, GTB, NAGAT Mouse Monoclonal Antibody [clone: 593CT3.2.2]

Supplier: Abcepta
AM2158A
89511-950EA 637.11 USD InStock
89511-950
Anti-A3GALNT, A3GALT1, GTB, NAGAT Mouse Monoclonal Antibody [clone: 593CT3.2.2]
Antibodies
Western Blot: 1:200~6400,

Type: Primary
Antigen: A3GALNT, A3GALT1, GTB, NAGAT
Clonality: Monoclonal
Clone: 593CT3.2.2
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty