Would you like to log in again?
DEV Nutrient Agar, MilliporeSigma
Supplier: MilliporeSigma
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
1.11471.0500
EM1.11471.0500EA 352.4 USD InStock
EM1.11471.0500
DEV Nutrient Agar, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
  • Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
  • Greater than or equal to 70% recovery on test medium Klebsiella pneumoniae ATCC 13883 (WDCM 00097)
  • Greater than or equal to 70% recovery on test medium Serratia marcescens ATCC 14756
  • Greater than or equal to 70% recovery on test medium Proteus hauseri ATCC 13315
  • Greater than or equal to 70% recovery on test medium Aeromonas hydrophila ATCC 7966 (WDCM 00063)
  • Greater than or equal to 70% recovery on test medium Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)
  • Greater than or equal to 70% recovery on test medium Enterococcus faecalis ATCC 11700
  • Greater than or equal to 70% recovery on test medium Bacillus subtilis ATCC 6633 (WDCM 00003)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty