Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrouridine, Calbiochem®

Supplier: MilliporeSigma
Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-hydroxytetrahydropyrimidin-2(1H)-one

584222-10MG 584222-10MG
80054-664EA 276.75 USD InStock
80054-664 CA80054-664
Tetrahydrouridine, Calbiochem®
Tetrahydrouridine
Tetrahydrouridine is a potent, competitive inhibitor of cytidine deaminase.

  • Competitive inhibitor of cytidine deaminase
  • Assess anti-leukemic activitiy of Ara-C
  • Used in in vitro studies

A potent competitive inhibitor of cytidine deaminase. Used in combination with cytosine arabinoside (Ara-C) to assess the anti-leukemic activity and anti-tumor activity of Ara-C in in vitro studies.


Formula: C₉H₁₆N₂O₆
MW: 248.2331 g/mol
MDL Number: MFCD00063468
CAS Number: 18771-50-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty