Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminopyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: Pyrimidin-2-ylamine

A0412-25G
TCA0412-25GEA 44.66 USD InStock
TCA0412-25G
2-Aminopyrimidine ≥98.0% (by GC, titration analysis)
2-Aminopyrimidine
CAS Number: 109-12-6
MDL Number: MFCD00006089
Molecular Formula: C4H5N3
Molecular Weight: 95.11
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Formula: C₄H₅N₃
MW: 95.1 g/mol
Melting Pt: 125 °C
MDL Number: MFCD00006089
CAS Number: 109-12-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/78/49/28927849.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty