Synonyms:
Aminobenzene Phenylamine
A0463-500G
TCA0463-500GEA
53.74
USD
InStock
TCA0463-500G
Aniline ≥98.0% (by GC, titration analysis)
Aniline
CAS Number: 62-53-3
MDL Number: MFCD00007629
Molecular Formula: C6H7N
Molecular Weight: 93.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 184
Melting point (°C): -6
Flash Point (°C): 75
Specific Gravity (20/20): 1.02