Synonyms:
Aluminumtert-butoxide
, Aluminium tert-butoxide
A0804-5G
TCA0804-005GEA
49.74
USD
InStock
TCA0804-005G
Aluminium tri-tert-butanolate ≥98.0% (by titrimetric analysis)
Aluminium tri-tert-butanolate
CAS Number: 556-91-2
MDL Number: MFCD00008801
Molecular Formula: C12H27AlO3
Molecular Weight: 246.33
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 180
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts
Other Product Names / Synonyms: Aluminum tert-Butylate