Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonia ∼4% ∼2.0 mol/L in methanol

Supplier: TCI America

A1884-500ML
TCA1884-500MLEA 142.68 USD InStock
TCA1884-500ML
Ammonia ∼4% ∼2.0 mol/L in methanol
Ammonia
CAS Number: 7664-41-7
MDL Number: MFCD00011418
Molecular Formula: H3N
Molecular Weight: 17.03
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 6
Storage Temperature: 0-10°C


Formula: NH₃
MW: 17.03 g/mol
Flash Pt: 6 °C
MDL Number: MFCD00011418
CAS Number: 7664-41-7
UN: 1005
ADR: 2,
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/70/28929970.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty