Synonyms:
2-Methylpyridin-4-amine
, 2-Methyl-4-pyridylamine
, 4-Amino-2-picoline
A2348-1G
TCA2348-1GEA
39.59
USD
InStock
TCA2348-1G
4-Amino-2-methylpyridine ≥97.0% (by GC)
4-Amino-2-methylpyridine
CAS Number: 18437-58-6
MDL Number: MFCD00186509
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 143
Melting point (°C): 96
Other Product Names / Synonyms: 4-Amino-2-picoline