Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-β-D-Arabinofuranosyluracil ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: Ara-UUracil 1-beta-D-Arabinofuranoside

A2356-5G A2356-1G
TCA2356-5GEA 38 USD InStock
TCA2356-5G TCA2356-1G
1-β-D-Arabinofuranosyluracil ≥98.0% (by HPLC, titration analysis)
1-beta-D-Arabinofuranosyluracil
CAS Number: 3083-77-0
MDL Number: MFCD00065998
Molecular Formula: C9H12N2O6
Molecular Weight: 244.20
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 222
Specific rotation [a]20/D: 120 deg (C=0.5, H2O)
Storage Temperature: 0°C

Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: Ara-U, Uracil 1-beta-D-Arabinofuranoside


Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00065998
CAS Number: 3083-77-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/46/28930546.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty