Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

11-Azido-3,6,9-trioxaundecan-1-amine ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 1-Amino-11-azido-3,6,9-trioxaundecane11-Amino-3,6,9-trioxaundecanyl AzideAmino-PEG3-AzideAzido-PEG3-Amine

A2363-1G A2363-5G
TCA2363-5GEA 1026.93 USD InStock
TCA2363-5G TCA2363-1G
11-Azido-3,6,9-trioxaundecan-1-amine ≥95.0% (by titrimetric analysis)
11-Azido-3,6,9-trioxaundecan-1-amine
CAS Number: 134179-38-7
MDL Number: MFCD00269874
Molecular Formula: C8H18N4O3
Molecular Weight: 218.26
Purity/Analysis Method: 95.0% (T)
Form: Clear Liquid
Specific Gravity (20/20): 1.10
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Azide Compounds (Click Chemistry), Bioscience, Crosslinkers, PEGylation Reagents & PEG Spacers
Other Product Names / Synonyms: 1-Amino-11-azido-3,6,9-trioxaundecane, 11-Amino-3,6,9-trioxaundecanyl Azide, Amino-PEG3-Azide, Azido-PEG3-Amine


Formula: C₈H₁₈N₄O₃
MW: 218.26 g/mol
Density: 1.10 (20 °C)
MDL Number: MFCD00269874
CAS Number: 134179-38-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/52/28930552.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty