Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥98.0% (by HPLC) for biochemical research

Supplier: TCI America
Synonyms: 2-(4-Amidinophenyl)indole-6-carboxamidine DihydrochlorideDAPI.2HCl2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride

A2412-5MG
TCA2412-5MGEA 84.56 USD InStock
TCA2412-5MG
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥98.0% (by HPLC) for biochemical research
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride)
[for Biochemical Research]
CAS Number: 28718-90-3
MDL Number: MFCD00012681
Molecular Formula: C16H15N5
Molecular Weight: 350.25
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Storage Temperature: 0°C

Product Categories: Bioscience, Stains & Dyes, Fluorescent Stains
Other Product Names / Synonyms: 4',6-Diamidino-2-phenylindole Dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride


Formula: C₁₆H₁₅N₅
MW: 350.25 g/mol
MDL Number: MFCD00012681
CAS Number: 28718-90-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/96/28930596.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty