Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-Amino-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: H-DL-β-Phe-OHβ-Aminohydrocinnamic acid3-Amino-3-phenylpropionic acidDL-β-Phenylalanine

A2480-5G A2480-25G
TCA2480-25GEA 59.89 USD InStock
TCA2480-25G TCA2480-5G
(±)-3-Amino-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)
(±)-3-Amino-3-phenylpropionic acid
CAS Number: 614-19-7
MDL Number: MFCD00008064
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 223

Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: beta-Phenylalanine


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 223 °C
MDL Number: MFCD00008064
CAS Number: 614-19-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/79/28930679.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty