Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: (S)-4-Amino-2-(Boc-amino)butyric Acid(S)-Nalpha-Boc-2,4-diaminobutyric AcidBoc-Dab-OH

A2515-5G A2515-1G
TCA2515-5GEA 346 USD InStock
TCA2515-5G TCA2515-1G
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid ≥98.0% (by titrimetric analysis)
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid
CAS Number: 25691-37-6
MDL Number: MFCD00236841
Molecular Formula: C9H18N2O4
Molecular Weight: 218.25
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 202
Specific rotation [a]20/D: -10 deg (C=2, AcOH)

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: (S)-4-Amino-2-(Boc-amino)butyric Acid, (S)-Nalpha-Boc-2,4-diaminobutyric Acid, Boc-Dab-OH


Formula: C₉H₁₈N₂O₄
MW: 218.25 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00236841
CAS Number: 25691-37-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/21/28930721.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty