Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥98.0% (by HPLC)

Supplier: TCI America
Synonyms: 5-Amino-1-(beta-D-ribofuranosyl)-1H-imidazole-4-carboxamideAcadesineAICAR

A2528-50MG
TCA2528-50MGEA 100.22 USD InStock
TCA2528-50MG
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside ≥98.0% (by HPLC)
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside
CAS Number: 2627-69-2
MDL Number: MFCD00869751
Molecular Formula: C9H14N4O5
Molecular Weight: 258.23
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 212
Specific rotation [a]20/D: -63 deg (C=1, H2O)
Lambda max.: 265 nm (NaOH)Storage Temperature: 0°C

Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents, Antitumor Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 5-Amino-1-(beta-D-ribofuranosyl)-1H-imidazole-4-carboxamide, Acadesine, AICAR


Formula: C₉H₁₄N₄O₅
MW: 258.23 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00869751
CAS Number: 2627-69-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/07/33/28930733.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty