Synonyms:
H-Lys(Ac)-OH
A2652-1G
TCA2652-1GEA
52.85
USD
InStock
TCA2652-1G
Nε-acetyl-L-lysine ≥98.0% (by titrimetric analysis)
Nepsilon-acetyl-L-lysine
CAS Number: 692-04-6
MDL Number: MFCD00002639
Molecular Formula: C8H16N2O3
Molecular Weight: 188.23
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 240
Specific rotation [a]20/D: 4 deg (C=1, H2O)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free), Biologically-modified Amino Acids
Other Product Names / Synonyms: H-Lys(Ac)-OH