Synonyms:
3-Azido-1-propanamine1-Amino-3-azidopropane3-Aminopropyl Azide
A2738-100MG
TCA2738-100MGEA
95.38
USD
InStock
TCA2738-100MG
3-Azidopropylamine ≥95.0% (by GC, titration analysis)
3-Azidopropylamine
CAS Number: 88192-19-2
MDL Number: MFCD11046568
Molecular Formula: C3H8N4
Molecular Weight: 100.13
Purity/Analysis Method: 95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 50
Flash Point (°C): 60
Specific Gravity (20/20): 1.02
Storage Temperature: 0°C
Product Categories: Synthetic Organic Chemistry, Azide Compounds (Click Chemistry), Bioscience, Crosslinkers
Other Product Names / Synonyms: 3-Azido-1-propanamine, 1-Amino-3-azidopropane, 3-Aminopropyl Azide